Anti-LTB4R2 Rabbit Polyclonal Antibody
Catalog # PRSI16-063
Supplier: ProSci Inc.
Specifications
- Antibody type:Primary
- Antigen name:Leukotriene B4 receptor 2
- Antigen symbol:LTB4R2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Western blot:Yes
- Form:Liquid
- Gene ID:56413
- Antigen synonyms:Leukotriene B4 receptor BLT2|Seven transmembrane receptor BLTR2|BLTR2|LTB4 receptor JULF2|LTB4-R 2|Leukotriene B4 receptor 2|LTB4-R2|BLT2R|LTB4R2
- Modification:None
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at −20 °C. Avoid freeze/thaw cycles
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 254-358 of human LTB4R2 (NP_062813.2)
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: LTB4R2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat