Anti-DYNLRB1 Rabbit Polyclonal Antibody
Catalog # PRSI15-923
Supplier: ProSci Inc.
Specifications
- Antibody type:Primary
- Antigen name:Dynein light chain roadblock-type 1
- Antigen symbol:DYNLRB1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:83658
- Antigen synonyms:DNLC2A|BITH|BLP|cytoplasmic|ROBL/LC7-like 1|dynein-associated protein Km23|roadblock domain-containing protein 1|ROBLD1|DNCL2A|dynein|light polypeptide 2A|bithoraxoid-like protein|dynein light chain roadblock-type 1
- Modification:None
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at −20 °C. Avoid freeze/thaw cycles
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:A synthetic peptide corresponding to a sequence within amino acids 1-63 of human DYNLRB1 (NP_001268656.1)
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: DYNLRB1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse