Anti-NOS3 Rabbit Polyclonal Antibody
PRSI15-861
- Antibody type:Primary
- Antigen name:Nitric oxide synthase 3 (endothelial cell)
- Antigen symbol:NOS3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Western blot:Yes
- Form:Liquid
- Gene ID:4846
- Antigen synonyms:Nitric oxide synthase 3|CNOS|Constitutive NOS|NOS type III|ENOS|NOSIII|ECNOS|NOS3|EC-NOS|Endothelial NOS
- Modification:None
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at −20 °C. Avoid freeze/thaw cycles
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:A synthetic peptide corresponding to a sequence within amino acids 680-780 of human eNOS (NP_000594.2)
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Type: Primary
Antigen: NOS3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse