Anti-YTHDC2 Rabbit Polyclonal Antibody
PRSI15-826
- Antibody type:Primary
- Antigen name:YTH domain containing 2
- Antigen symbol:YTHDC2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Western blot:Yes
- Form:Liquid
- Gene ID:64848
- Antigen synonyms:CAHL|probable ATP-dependent RNA helicase YTHDC2|CsA-associated helicase-like protein|3'-5' RNA helicase YTHDC2|hYTHDC2
- Modification:None
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at −20 °C. Avoid freeze/thaw cycles
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 1131-1430 of human YTHDC2 (NP_073739.3)
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Type: Primary
Antigen: YTHDC2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse