Anti-RALGPS1 Rabbit Polyclonal Antibody
Catalog # PRSI15-751
Supplier: ProSci Inc.
Specifications
- Antibody type:Primary
- Antigen name:Ral GEF with PH domain and SH3 binding motif 1
- Antigen symbol:RALGPS1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Rat
- Western blot:Yes
- Form:Liquid
- Gene ID:9649
- Antigen synonyms:RALGPS1|RALGEF2A|ral guanine nucleotide exchange factor 2|Ral guanine nucleotide exchange factor RalGPS1A|ras-specific guanine nucleotide-releasing factor RalGPS1|ralA exchange factor RalGPS1
- Modification:None
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at −20 °C. Avoid freeze/thaw cycles
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 1-300 of human RALGPS1 (NP_055451.1)
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: RALGPS1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat