Anti-S100P Rabbit Polyclonal Antibody
Catalog # PRSI15-726
Supplier: ProSci Inc.
Specifications
- Antibody type:Primary
- Antigen name:S100 calcium binding protein P
- Antigen symbol:S100P
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Western blot:Yes
- Form:Liquid
- Gene ID:6286
- Antigen synonyms:protein S100-P|migration-inducing gene 9 protein|MIG9|protein S100-E
- Modification:None
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at −20 °C. Avoid freeze/thaw cycles
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:A synthetic peptide corresponding to a sequence within amino acids 1-95 of human S100P (NP_005971.1)
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: S100P
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat