Anti-BCAM Rabbit Polyclonal Antibody
PRSI15-702
- Antibody type:Primary
- Antigen name:Basal cell adhesion molecule (Lutheran blood group)
- Antigen symbol:BCAM
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:4059
- Antigen synonyms:MSK19|F8/G253 antigen|B-cell adhesion molecule|CD239|AU|B-CAM cell surface glycoprotein|Auberger b antigen|Lutheran blood group variant LUGA|basal cell adhesion molecule (Lu and Au blood groups)|LU|basal cell adhesion molecule
- Modification:None
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at −20 °C. Avoid freeze/thaw cycles
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 352-547 of human BCAM (NP_005572.2)
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Type: Primary
Antigen: BCAM
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse