Anti-DYNC1I2 Rabbit Polyclonal Antibody
PRSI15-686
New Product
- Antibody type:Primary
- Antigen name:Dynein cytoplasmic 1 intermediate chain 2
- Antigen symbol:DYNC1I2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Western blot:Yes
- Form:Liquid
- Gene ID:1781
- Antigen synonyms:IC2|intermediate polypeptide 2|testis tissue sperm-binding protein Li 66n|cytoplasmic|cytoplasmic dynein 1 intermediate chain 2|DNCI2|cytosolic|DH IC-2|dynein|DIC74|dynein intermediate chain 2
- Modification:None
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at −20 °C. Avoid freeze/thaw cycles
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 352-612 of human DYNC1I2 (NP_001258717.1)
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Type: Primary
Antigen: DYNC1I2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat