Anti-CA5A Rabbit Polyclonal Antibody
Catalog # PRSI15-678
Supplier: ProSci Inc.
New Product
Specifications
- Antibody type:Primary
- Antigen name:Carbonic anhydrase 5A
- Antigen symbol:CA5A
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Form:Liquid
- Gene ID:763
- Antigen synonyms:CAV|carbonic dehydratase|CA5D|CA5A|carbonic anhydrase VA|carbonate dehydratase VA|carbonic anhydrase V|CA-VA|GS1-21A4.1|carbonic anhydrase 5A|mitochondrial|CAVA
- Modification:None
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at −20 °C. Avoid freeze/thaw cycles
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 39-305 of human CA5A (NP_001730.1)
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: CA5A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human