Anti-FOSB Rabbit Polyclonal Antibody
Catalog # PRSI15-671
Supplier: ProSci Inc.
Specifications
- Antikörper-Typ:Primär
- Antigen-Name:FBJ murine osteosarcoma viral oncogene homolog B
- Antigen-Symbol:FOSB
- Klonalität:Polyklonal
- Konjugation:Unconjugated
- Wirt:Rabbit
- Isotyp:IgG
- Reaktivität:Human,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Gen-ID:2354
- Antigen-Synonyme:AP-1|GOSB|G0S3|GOS3|FOSB
- Modification:None
- Lagerungspuffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Lagertemperatur:Store at −20 °C. Avoid freeze/thaw cycles
- Konzentration:Batch dependent
- Transporttemperatur:20
- Immunogen:A synthetic peptide corresponding to a sequence within amino acids 1-100 of human FOSB (NP_006723.2)
- Reinigung:Affinity purification
- Packung:Vial
- VE:50 µl
Specifications
About this item
Type: Primary
Antigen: FOSB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat