Anti-NOL6 Rabbit Polyclonal Antibody
Catalog # PRSI15-530
Supplier: ProSci Inc.
Specifications
- Antibody type:Primary
- Antigen name:Nucleolar protein 6 (RNA-associated)
- Antigen symbol:NOL6
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:65083
- Antigen synonyms:NRAP|MGC14896|FLJ21959|UTP22|MGC20838|bA311H10.1|MGC14921|NOL6
- Modification:None
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at −20 °C. Avoid freeze/thaw cycles
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 1032-1146 of human NOL6 (NP_075068.2)
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: NOL6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat