Anti-NDUFB3 Rabbit Polyclonal Antibody
Catalog # PRSI15-512
Supplier: ProSci Inc.
Specifications
- Antibody type:Primary
- Antigen name:NADH:ubiquinone oxidoreductase subunit B3
- Antigen symbol:NDUFB3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Western blot:Yes
- Form:Liquid
- Gene ID:4709
- Antigen synonyms:complex I-B12|NADH dehydrogenase (ubiquinone) 1 beta subcomplex|NADH dehydrogenase [ubiquinone] 1 beta subcomplex subunit 3|12kDa|B12|CI-B12|NADH-ubiquinone oxidoreductase B12 subunit|3|MC1DN25
- Modification:None
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at −20 °C. Avoid freeze/thaw cycles
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 1-98 of human NDUFB3 (NP_002482.1)
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: NDUFB3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse