Anti-SLC4A10 Rabbit Polyclonal Antibody
PRSI15-213
New Product
- Antibody type:Primary
- Antigen name:Solute carrier family 4 member 10
- Antigen symbol:SLC4A10
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Western blot:Yes
- Form:Liquid
- Gene ID:57282
- Antigen synonyms:NCBE|sodium bicarbonate transporter|NBCn2|sodium bicarbonate cotransporter-like|member 1solute carrier family 4|solute carrier family 4|sodium-driven chloride bicarbonate exchanger|member 10
- Modification:None
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at −20 °C. Avoid freeze/thaw cycles
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 640-725 of human SLC4A10 (NP_001171486.1)
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Type: Primary
Antigen: SLC4A10
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse