Anti-NF2 Rabbit Polyclonal Antibody
Catalog # PRSI15-108
Supplier: ProSci Inc.
Specifications
- Antibody type:Primary
- Antigen name:Neurofibromin 2 (merlin)
- Antigen symbol:NF2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Western blot:Yes
- Form:Liquid
- Gene ID:4771
- Antigen synonyms:Schwannomerlin|NF2|SCH|Merlin|ACN|BANF|Neurofibromin 2 (merlin)|Neurofibromin-2|Schwannomin|Moesin-ezrin-radixin like
- Modification:None
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at −20 °C. Avoid freeze/thaw cycles
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 477-576 of human NF2 (NP_000259.1)
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: NF2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat