Anti-GSTM2 Rabbit Polyclonal Antibody
Catalog # PRSI15-023
Supplier: ProSci Inc.
Specifications
- Antibody type:Primary
- Antigen name:Glutathione S-transferase mu 2 (muscle)
- Antigen symbol:GSTM2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Western blot:Yes
- Form:Liquid
- Gene ID:2946
- Antigen synonyms:GSTM2|GTHMUS|MGC117303|GSTM2-2|GST4|GSTM
- Modification:None
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at −20 °C. Avoid freeze/thaw cycles
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 1-218 of human GSTM2 (NP_000839.1)
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: GSTM2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse