Anti-Cd5 Rat Monoclonal Antibody [clone: B19.1] (PE (Phycoerythrin))
Catalog # PRSI98-599
Supplier: ProSci Inc.
Specifications
- Antibody type:Primary
- Antigen name:CD5 antigen
- Antigen symbol:Cd5
- Clonality:Monoclonal
- Clone:[B19.1]
- Conjugation:PE (Phycoerythrin)
- ELISA:Yes
- Flow cytometry:Yes
- Host:Rat
- Isotype:IgG2a
- Reactivity:Mouse
- Form:Liquid
- Gene ID:12507
- Antigen synonyms:Ly-A|Ly-12|Tp67|Lymphocyte antigen 1|Ly-1|T1|Lyt-1
- Modification:None
- Storage buffer:Supplied in PBS/NaN₃ and a stabilising agent
- Storage temperature:Store vial at 2...8 °C
- Concentration:0,1 mg/ml
- Shipping temperature:20
- Pack type:Vial
- Pk:0,1 mg
Specifications
About this item
Type: Primary
Antigen: Cd5
Clonality: Monoclonal
Clone: [B19.1]
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse