Anti-Cd4 Rat Monoclonal Antibody [clone: GK1.5] (APC (Allophycocyanin)-Cy7®)
Catalog # PRSI98-595
Supplier: ProSci Inc.
Specifications
- Antibody type:Primary
- Antigen name:CD4 antigen
- Antigen symbol:Cd4
- Clonality:Monoclonal
- Clone:[GK1.5]
- Conjugation:APC (Allophycocyanin)-Cy7®
- ELISA:Yes
- Flow cytometry:Yes
- Host:Rat
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG2b
- Western blot:Yes
- Form:Liquid
- Gene ID:12504
- Antigen synonyms:L3T4|Ly-4
- Modification:None
- Storage buffer:Supplied in PBS/NaN₃ and a stabilising agent
- Storage temperature:Store vial at 2...8 °C
- Concentration:0,1 mg/ml
- Shipping temperature:20
- Immunogen:Mouse CTL clone V4
- Tested applications:Multiplex
- Pack type:Vial
- Pk:0,1 mg
Specifications
About this item
Type: Primary
Antigen: Cd4
Clonality: Monoclonal
Clone: [GK1.5]
Conjugation: APC (Allophycocyanin)-Cy7®
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse