Anti-Cd3e Rat Monoclonal Antibody [clone: C363.29B] (PE (Phycoerythrin))
Catalog # PRSI98-578
Supplier: ProSci Inc.
Specifications
- Antibody type:Primary
- Antigen name:CD3 antigen, epsilon polypeptide
- Antigen symbol:Cd3e
- Clonality:Monoclonal
- Clone:[C363.29B]
- Conjugation:PE (Phycoerythrin)
- ELISA:Yes
- Host:Rat
- Isotype:IgG2b
- Reactivity:Mouse
- Form:Liquid
- Gene ID:12501
- Antigen synonyms:CD3 epsilon
- Modification:None
- Storage buffer:Supplied in PBS/NaN₃ and a stabilising agent
- Storage temperature:Store vial at 2...8 °C
- Concentration:0,1 mg/ml
- Shipping temperature:20
- Immunogen:IL-4 producing Th2 cell lines including D10
- Pack type:Vial
- Pk:0,1 mg
Specifications
About this item
Type: Primary
Antigen: Cd3e
Clonality: Monoclonal
Clone: [C363.29B]
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse