Order Entry
Switzerland
ContactUsLinkComponent
Anti-ICAM1 Rabbit Monoclonal Antibody [clone: H3]
Anti-ICAM1 Rabbit Monoclonal Antibody [clone: H3]
Catalog # PRSI54-324
Supplier:  ProSci Inc.
Anti-ICAM1 Rabbit Monoclonal Antibody [clone: H3]
Catalog # PRSI54-324
Supplier:  ProSci Inc.

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    intercellular adhesion molecule 1
  • Antigen symbol:
    ICAM1
  • Clonality:
    Monoclonal
  • Clone:
    H3
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Western blot:
    Yes
  • Gene ID:
    3383
  • Antigen synonyms:
    BB2|CD54|P3.58
  • Modification:
    None
  • Storage buffer:
    PBS, pH 7,2, 0,02% NaN₃
  • Storage temperature:
    Store at 4 °C for up to 6 months. Store at −20 °C long term
  • Concentration:
    Batch dependent
  • Shipping temperature:
    20
  • Immunogen:
    Synthetic peptide derived from human intercellular adhesion molecule 1 (ICAM1)(ASVSVTAEDEGTQR)
  • Purification:
    Protein A purified
  • Pack type:
    Vial
  • Pk:
    100 µG

Specifications

About this item

Type: Primary
Antigen: ICAM1
Clonality: Monoclonal
Clone: H3
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human