Order Entry
Switzerland
ContactUsLinkComponent
Anti-ENO1 Rabbit Monoclonal Antibody [clone: A6]
Anti-ENO1 Rabbit Monoclonal Antibody [clone: A6]
Catalog # PRSI54-318
Supplier:  ProSci Inc.
Anti-ENO1 Rabbit Monoclonal Antibody [clone: A6]
Catalog # PRSI54-318
Supplier:  ProSci Inc.

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    enolase 1
  • Antigen symbol:
    ENO1
  • Clonality:
    Monoclonal
  • Clone:
    A6
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
    IgG kappa
  • Western blot:
    Yes
  • Gene ID:
    2023
  • Antigen synonyms:
    ENO1L1|NNE|HEL-S-17|PPH|MPB1
  • Modification:
    None
  • Storage buffer:
    49% PBS, 50% Glycerol, 0,05% BSA, pH 7,2, 0,02% NaN₃
  • Storage temperature:
    Store at 4 °C for up to 6 months. Store at −20 °C long term
  • Concentration:
    Batch dependent
  • Shipping temperature:
    20
  • Immunogen:
    Synthetic peptides derived from human ENOA/ENO1 (DATNVGDEGGFAPNILENK)
  • Purification:
    Protein A purified
  • Pack type:
    Vial
  • Pk:
    100 µG

Specifications

About this item

Type: Primary
Antigen: ENO1
Clonality: Monoclonal
Clone: A6
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG kappa
Reactivity: Human, Mouse