Anti-IFITM3 Rabbit Polyclonal Antibody
Catalog # PRSI14-777
Supplier: ProSci Inc.
Specifications
- Antibody type:Primary
- Antigen name:Interferon-induced transmembrane protein 3
- Antigen symbol:IFITM3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat
- Western blot:Yes
- Form:Liquid
- Gene ID:10410
- Antigen synonyms:Dispanin subfamily A member 2b|DSPA2b|Interferon-induced transmembrane protein 3|IFITM3|Interferon-inducible protein 1-8U
- Modification:None
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at –20...+730 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 1-133 of human IFITM3 (NP_066362.2)
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: IFITM3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat