Anti-SLC6A11 Rabbit Polyclonal Antibody
Codice catalogo PRSI14-106
Nome del fornitore: ProSci Inc.
Specifiche
- Antibody type:Primary
- Antigen name:solute carrier family 6 member 11
- Antigen symbol:SLC6A11
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:6538
- Antigen synonyms:member 11|GAT4|GABA transporter 3|solute carrier family 6 (neurotransmitter transporter)|GABA)|GAT3|GAT-3|sodium- and chloride-dependent GABA transporter 3|solute carrier family 6 (neurotransmitter transporter
- Modification:None
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at –20...+730 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 1-60 of human SLC6A11 (NP_055044.1)
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Specifiche
Informazioni su questo articolo
Type: Primary
Antigen: SLC6A11
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat