Order Entry
Switzerland
ContactUsLinkComponent
Anti-RAB11A Rabbit Polyclonal Antibody
Anti-RAB11A Rabbit Polyclonal Antibody
Catalog # PRSI14-092
Supplier:  ProSci Inc.
Anti-RAB11A Rabbit Polyclonal Antibody
Catalog # PRSI14-092
Supplier:  ProSci Inc.

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    RAB11A, member RAS oncogene family
  • Antigen symbol:
    RAB11A
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Western blot:
    Yes
  • Form:
    Liquid
  • Gene ID:
    8766
  • Antigen synonyms:
    RAB11A|YL8|RAB 11A|member oncogene family|RAB11|member RAS oncogene family
  • Modification:
    None
  • Storage buffer:
    PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
  • Storage temperature:
    Store at –20...+730 °C. Avoid freeze/thaw cycles.
  • Concentration:
    Batch dependent
  • Shipping temperature:
    20
  • Immunogen:
    Recombinant fusion protein containing a sequence corresponding to amino acids 1-216 of human RAB11A (NP_004654.1)
  • Purification:
    Affinity purification
  • Pack type:
    Vial
  • Pk:
    50 µl

Specifications

About this item

Type: Primary
Antigen: RAB11A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat