Anti-SLC6A5 Rabbit Polyclonal Antibody
Catalog # PRSI14-082
Supplier: ProSci Inc.
Specifications
- Antibody type:Primary
- Antigen name:solute carrier family 6 member 5
- Antigen symbol:SLC6A5
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Western blot:Yes
- Form:Liquid
- Gene ID:9152
- Antigen synonyms:sodium- and chloride-dependent glycine transporter 2|HKPX3|GLYT-2|GLYT2|NET1|glycine)|member 5|solute carrier family 6 (neurotransmitter transporter
- Modification:None
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at –20...+730 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 1-200 of human SLC6A5 (NP_004202.3)
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: SLC6A5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat