Anti-NT5C2 Rabbit Polyclonal Antibody
Catalog # PRSI14-073
Supplier: ProSci Inc.
Specifications
- Antibody type:Primary
- Antigen name:5'-nucleotidase, cytosolic II
- Antigen symbol:NT5C2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:22978
- Antigen synonyms:epididymis secretory sperm binding protein|cytosolic purine 5'-nucleotidase|PNT5|SPG45|GMP|NT5B|SPG65|cytosolic type B|5'-nucleotidase (purine)|cN-II|IMP-specific 5'-NT
- Modification:None
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at –20...+730 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 382-561 of human NT5C2 (NP_036361.1)
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: NT5C2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat