Anti-PDE1A Rabbit Polyclonal Antibody
Catalog # PRSI13-662
Supplier: ProSci Inc.
Specifications
- Antibody type:Primary
- Antigen name:phosphodiesterase 1A, calmodulin-dependent
- Antigen symbol:PDE1A
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Western blot:Yes
- Form:Liquid
- Gene ID:5136
- Antigen synonyms:calcium/calmodulin-stimulated cyclic nucleotide phosphodiesterase|phosphodiesterase-1A|3'|CAM-PDE-1A|MGC26303|calmodulin-dependent|HSPDE1A|PDE1A|HCAM1|calmodulin-dependent phosphodiesterase|5' cyclic nucleotide phosphodiesterase|phosphodiesterase 1A|HCAM-1
- Modification:None
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at –20...+730 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 446-545 of human PDE1A (NP_005010.2)
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: PDE1A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat