Anti-NPR1 Rabbit Polyclonal Antibody
PRSI13-654
New Product
- Antibody type:Primary
- Antigen name:natriuretic peptide receptor 1
- Antigen symbol:NPR1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Western blot:Yes
- Form:Liquid
- Gene ID:4881
- Antigen synonyms:natriuretic peptide receptor A|atrial natriuretic peptide receptor 1|natriuretic peptide A type receptor|ANPa|ANPRA|NPR-A|NPRA|atrial natriuretic peptide receptor type A|testicular tissue protein Li 20|GC-A|ANP-A|GUCY2A|GUC2A|guanylate cyclase A|atrionatriuretic peptide receptor A|ANPR-A|natriuretic peptide receptor A/guanylate cyclase A (atrionatriuretic peptide receptor A)
- Modification:None
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at –20...+730 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 260-470 of human NPR1 (NP_000897.3)
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Type: Primary
Antigen: NPR1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat