Anti-S100A14 Rabbit Polyclonal Antibody
Catalog # PRSI13-624
Supplier: ProSci Inc.
New Product
Specifications
- Antibody type:Primary
- Antigen name:S100 calcium binding protein A14
- Antigen symbol:S100A14
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Form:Liquid
- Gene ID:57402
- Antigen synonyms:S114|S100A15|breast cancer membrane protein 84|protein S100-A14|BCMP84
- Modification:None
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at –20...+730 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 1-104 of human S100A14 (NP_065723.1)
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: S100A14
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat