Anti-SNAPIN Rabbit Polyclonal Antibody
PRSI13-562
- Antibody type:Primary
- Antigen name:SNAP-associated protein
- Antigen symbol:SNAPIN
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:23557
- Antigen synonyms:BLOC1S7|SNAREassociated protein Snapin|SNARE-associated protein Snapin|SNAP-associated protein|BLOS7|BLOC-1 subunit 7|SNAP25BP|Biogenesis of lysosome-related organelles complex 1 subunit 7|SNAPAP|Synaptosomal-associated protein 25 binding protein
- Modification:None
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at –20...+730 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 1-136 of human SNAPIN (NP_036569.1)
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Type: Primary
Antigen: SNAPIN
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat