Anti-SGTA Rabbit Polyclonal Antibody
Catalog # PRSI22-851
Supplier: ProSci Inc.
Specifications
- Antibody type:Primary
- Antigen name:Small glutamine-rich tetratricopeptide repeat-containing protein alpha
- Antigen symbol:SGTA
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Form:Liquid
- Gene ID:6449
- Antigen synonyms:Alpha-SGT|UBP|Small glutamine-rich tetratricopeptide repeat-containing protein alpha|SGT|SGT1|Vpu-binding protein|SGTA
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at −20...+730 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 1-120 of human SGTA (NP_003012.1)
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: SGTA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human