Anti-GRM4 Rabbit Polyclonal Antibody
Catalog # PRSI22-834
Supplier: ProSci Inc.
Specifications
- Antibody type:Primary
- Antigen name:Glutamate metabotropic receptor 4
- Antigen symbol:GRM4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Western blot:Yes
- Form:Liquid
- Gene ID:2914
- Antigen synonyms:GPRC1D|MGLUR4|glutamate receptor|metabotropic glutamate receptor 4|metabotropic 4|mGlu4
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at −20...+730 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 33-290 of human GRM4 (NP_000832.1)
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: GRM4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat