Anti-TES Rabbit Polyclonal Antibody
Katalognr. PRSI22-692
Anbieter: ProSci Inc.
Technische Daten
- Antibody type:Primary
- Antigen name:Testin
- Antigen symbol:TES
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat
- Western blot:Yes
- Form:Liquid
- Gene ID:26136
- Antigen synonyms:TES|TESS|Testin
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at −20...+730 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 1-260 of human TES (NP_056456.1)
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Technische Daten
Über diesen Artikel
Type: Primary
Antigen: TES
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat