Anti-TCEB3 Rabbit Polyclonal Antibody
Catalog # PRSI22-616
Supplier: ProSci Inc.
Specifications
- Antibody type:Primary
- Antigen name:Transcription elongation factor B (SIII), polypeptide 3 (110kDa, elongin A)
- Antigen symbol:TCEB3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:6924
- Antigen synonyms:SIII p110|SIII|TCEB3|EloA|TCEB3A
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at −20...+730 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 679-798 of human TCEB3 (NP_003189.2)
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: TCEB3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat