Anti-SSBP1 Rabbit Polyclonal Antibody
Catalog # PRSI22-609
Supplier: ProSci Inc.
Specifications
- Antibody type:Primary
- Antigen name:Single stranded DNA binding protein 1
- Antigen symbol:SSBP1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Western blot:Yes
- Form:Liquid
- Gene ID:6742
- Antigen synonyms:Mt-SSB|mitochondrial|SOSS-B1|SSBP|single-stranded DNA binding protein 1|single-stranded DNA-binding protein|mtSSB|PWP1-interacting protein 17
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at −20...+730 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 1-148 of human SSBP1 (NP_001243442.1)
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: SSBP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse