Anti-CCL8 Rabbit Polyclonal Antibody
Catalog # PRSI22-600
Supplier: ProSci Inc.
Specifications
- Antibody type:Primary
- Antigen name:Chemokine (C-C motif) ligand 8
- Antigen symbol:CCL8
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Western blot:Yes
- Form:Liquid
- Gene ID:6355
- Antigen synonyms:HC14|SCYA8|MCP2|MCP-2|SCYA10|CCL8
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at −20...+730 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 23-99 of human CCL8 (NP_005614.2)
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: CCL8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse