Anti-HLA-DMA Rabbit Polyclonal Antibody
Codice catalogo PRSI22-557
Nome del fornitore: ProSci Inc.
Specifiche
- Antibody type:Primary
- Antigen name:Major histocompatibility complex, class II, DM alpha
- Antigen symbol:HLA-DMA
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Western blot:Yes
- Form:Liquid
- Gene ID:3108
- Antigen synonyms:really interesting new gene 6 protein|DMA|M alpha chain|D6S222E|MHC class II antigen DMA|class II histocompatibility antigen|HLA class II histocompatibility antigen|RING6|HLADM|DM alpha chain
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at −20...+730 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 27-233 of human HLA-DMA (NP_006111.2)
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Specifiche
Informazioni su questo articolo
Type: Primary
Antigen: HLA-DMA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse