Anti-EPHX2 Rabbit Polyclonal Antibody
Catalog # PRSI18-364
Supplier: ProSci Inc.
Specifications
- Antibody type:Primary
- Antigen name:Epoxide hydrolase 2, cytoplasmic
- Antigen symbol:EPHX2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:2053
- Antigen synonyms:EPHX2|CEH|bifunctional epoxide hydrolase 2|SEH|cytoplasmic|cytosolic|soluble|epoxide hydrolase|epoxide hydratase|epoxide hydrolase 2
- Modification:None
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant protein of human EPHX2
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: EPHX2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat