Anti-GNB1 Rabbit Polyclonal Antibody
Catalog # PRSI18-356
Supplier: ProSci Inc.
New Product
Specifications
- Antibody type:Primary
- Antigen name:G protein subunit beta 1
- Antigen symbol:GNB1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:2782
- Antigen synonyms:transducin beta chain 1|guanine nucleotide-binding protein G(I)/G(S)/G(T) subunit beta-1|signal-transducing proteins GS/GI|guanine nucleotide binding protein (G protein)|MRD42|testicular tissue protein Li 72|beta polypeptide 1|beta subunit
- Modification:None
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 1-340 of human GNB1 (NP_002065,1)
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: GNB1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat