Anti-LGALS13 Rabbit Polyclonal Antibody
PRSI18-048
New Product
- Antibody type:Primary
- Antigen name:Galectin 13
- Antigen symbol:LGALS13
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:29124
- Antigen synonyms:gal-13|PLAC8|beta-galactoside-binding lectin|PP13|galactoside-binding|soluble|galactoside-binding soluble lectin 13|13|placental protein 13|GAL13|placental tissue protein 13|lectin
- Modification:None
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 30-139 of human LGALS13 (NP_037400,1)
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Type: Primary
Antigen: LGALS13
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat