Anti-BTRC Rabbit Polyclonal Antibody
PRSI16-657
New Product
- Antibody type:Primary
- Antigen name:Beta-transducin repeat containing E3 ubiquitin protein ligase
- Antigen symbol:BTRC
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:8945
- Antigen synonyms:FWD1|epididymis tissue protein Li 2a|F-box/WD repeat-containing protein 1A|betaTrCP|bTrCP1|E3RSIkappaB|bTrCP|BETA-TRCP|pIkappaBalpha-E3 receptor subunit|beta-TrCP1|FBXW1A|F-box and WD-repeat protein 1B|F-box and WD repeats protein beta-TrCP|FBXW1|FBW1A
- Modification:None
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 50-340 of human Beta-TRCP (NP_378663,1)
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Type: Primary
Antigen: BTRC
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat