Anti-IMPA1 Rabbit Polyclonal Antibody
PRSI22-174
New Product
- Antikörper-Typ:Primär
- Antigen-Name:Inositol monophosphatase 1
- Antigen-Symbol:IMPA1
- Klonalität:Polyklonal
- Konjugation:Unconjugated
- Wirt:Rabbit
- ImmunoChemie:Yes
- ImmunFluoreszenz:Yes
- Isotyp:IgG
- Western blot:Yes
- Form:Liquid
- Gen-ID:3612
- Antigen-Synonyme:lithium-sensitive myo-inositol monophosphatase A1|inositol(myo)-1(or 4)-monophosphatase 1|IMPA|D-galactose 1-phosphate phosphatase|IMP|IMPase 1|myo-inositol monophosphatase 1|inositol monophosphatase 1|testicular tissue protein Li 94|inositol-1(or 4)-monophosphatase 1|IMP 1|MRT59
- Lagerungspuffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3.
- Lagertemperatur:Store at −20...+730 °C. Avoid freeze/thaw cycles.
- Konzentration:Batch dependent
- Transporttemperatur:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 1-277 of human IMPA1 (NP_005527.1).
- Reinigung:Affinity purification
- Packung:Vial
- VE:50 µl
Type: Primary
Antigen: IMPA1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat