Anti-ZC3HC1 Rabbit Polyclonal Antibody
Catalog # PRSI22-134
Supplier: ProSci Inc.
Specifications
- Antibody type:Primary
- Antigen name:Zinc finger C3HC-type containing 1
- Antigen symbol:ZC3HC1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Western blot:Yes
- Form:Liquid
- Gene ID:51530
- Antigen synonyms:NIPA|nuclear-interacting partner of ALK|nuclear interacting partner of anaplastic lymphoma kinase (ALK)|hematopoietic stem/progenitor cell protein 216|HSPC216
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3.
- Storage temperature:Store at −20...+730 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 1-260 of human ZC3HC1 (NP_057562.3).
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: ZC3HC1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse