Anti-ZMYND11 Rabbit Polyclonal Antibody
PRSI22-130
New Product
- Antikörper-Typ:Primär
- Antigen-Name:Zinc finger, MYND-type containing 11
- Antigen-Symbol:ZMYND11
- Klonalität:Polyklonal
- Konjugation:Unconjugated
- Wirt:Rabbit
- Isotyp:IgG
- Reaktivität:Human,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Gen-ID:10771
- Antigen-Synonyme:RP11-486H9.1|MGC111056|BRAM1|BS69|ZMYND11
- Lagerungspuffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3.
- Lagertemperatur:Store at −20...+730 °C. Avoid freeze/thaw cycles.
- Konzentration:Batch dependent
- Transporttemperatur:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 278-567 of human ZMYND11 (NP_997644.2).
- Reinigung:Affinity purification
- Packung:Vial
- VE:50 µl
Type: Primary
Antigen: ZMYND11
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat