Anti-ANXA4 Rabbit Polyclonal Antibody
Catalog # PRSI22-099
Supplier: ProSci Inc.
Specifications
- Antibody type:Primary
- Antigen name:Annexin A4
- Antigen symbol:ANXA4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:307
- Antigen synonyms:DKFZp686H02120|ANX4|PIG28|ZAP36|MGC75105|ANXA4|HEL-S-274
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3.
- Storage temperature:Store at −20...+730 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 112-321 of human ANXA4 (NP_001144.1).
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: ANXA4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat