Anti-SLC7A5 Rabbit Polyclonal Antibody
Catalog # PRSI18-830
Supplier: ProSci Inc.
New Product
Specifications
- Antibody type:Primary
- Antigen name:Solute carrier family 7 (amino acid transporter light chain, L system), member 5
- Antigen symbol:SLC7A5
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Western blot:Yes
- Form:Liquid
- Gene ID:8140
- Antigen synonyms:large neutral amino acids transporter 1|large neutral amino acids transporter small subunit 1|E16|SLC7A5|y+ system)|CD98 light chain|4F2LC|CD98LC|hLAT1|LAT1|sodium-independent neutral amino acid transporter LAT1|CD98|MPE16|D16S469E|solute carrier family 7 (cationic amino acid transporter|member 5|4F2 light chain|integral membrane protein E16|L-type amino acid transporter 1
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3.
- Storage temperature:Store at −20...+730 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 1-125 of human SLC7A5 (NP_003477.4).
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: SLC7A5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat