Anti-PSMD3 Rabbit Polyclonal Antibody
Catalog # PRSI18-814
Anbieter: ProSci Inc.
New Product
Technische Daten
- Antibody type:Primary
- Antigen name:Proteasome 26S subunit, non-ATPase 3
- Antigen symbol:PSMD3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Western blot:Yes
- Form:Liquid
- Gene ID:5709
- Antigen synonyms:26S proteasome regulatory subunit S3|26S proteasome regulatory subunit RPN3|non-ATPase|26S proteasome non-ATPase regulatory subunit 3|RPN3|proteasome subunit p58|proteasome (prosome|tissue specific transplantation antigen 2|TSTA2|macropain) 26S subunit|S3|3|P58
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3.
- Storage temperature:Store at −20...+730 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:A synthetic peptide of human PSMD3.
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Technische Daten
Über diesen Artikel
Type: Primary
Antigen: PSMD3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat