Anti-MRPL48 Rabbit Polyclonal Antibody
PRSI60-090
New Product
- Antibody type:Primary
- Antigen name:39S ribosomal protein L48, mitochondrial
- Antigen symbol:MRPL48
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:31 - 59 aa
- Form:Liquid
- Gene ID:51642
- Modification:None
- Molecular weight:24 kDa
- Storage temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:0,5 mg/ml
- Shipping temperature:4°C
- Immunogen:This MRPL48 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 31 - 59 amino acids from the N-terminal region of human MRPL48.
- Purification:This antibody is purified through a protein A column, followed by peptide affinity purification
- Pk:400 µl
For WB starting dilution is: 1:1000.
Type: Primary
Antigen: MRPL48
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 31-59
Host: Rabbit
Isotype: Ig
Reactivity: Human