Anti-EAF1 Rabbit Polyclonal Antibody
Catalog # PRSI60-020
Supplier: ProSci Inc.
New Product
Specifications
- Antikörper-Typ:Primär
- Antigen-Name:ELL-associated factor 1
- Antigen-Symbol:EAF1
- Klonalität:Polyklonal
- Konjugation:Unconjugated
- Wirt:Rabbit
- Reaktivität:Human
- Western blot:Yes
- Epitop:58 - 85 aa
- Form:Liquid
- Gen-ID:85403
- Modification:None
- Molecular weight:29 kDa
- Lagertemperatur:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Konzentration:0,5 mg/ml
- Transporttemperatur:4°C
- Immunogen:This EAF1 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 58 - 85 amino acids from the Central region of human EAF1.
- Reinigung:This antibody is purified through a protein A column, followed by peptide affinity purification
- VE:400 µl
Specifications
About this item
For WB starting dilution is: 1:1000.
Type: Primary
Antigen: EAF1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 58-85
Host: Rabbit
Isotype: Ig
Reactivity: Human