Order Entry
Switzerland
ContactUsLinkComponent
Anti-ZNF484 Rabbit Polyclonal Antibody
Anti-ZNF484 Rabbit Polyclonal Antibody
Catalog # PRSI59-830
Supplier:  ProSci Inc.
New Product
undefined
Anti-ZNF484 Rabbit Polyclonal Antibody
Catalog # PRSI59-830
Supplier:  ProSci Inc.
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Zinc finger protein 484
  • Antigen symbol:
    ZNF484
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Epitope:
    100 - 127 aa
  • Form:
    Liquid
  • Gene ID:
    83744
  • Modification:
    None
  • Molecular weight:
    98 kDa
  • Storage temperature:
    Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
  • Concentration:
    0,32 mg/ml
  • Shipping temperature:
    4°C
  • Immunogen:
    This ZNF484 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 100 - 127 amino acids from the N-terminal region of human ZNF484.
  • Purification:
    This antibody is purified through a protein A column, followed by peptide affinity purification
  • Pk:
    400 µl

Specifications

About this item

For WB starting dilution is: 1:1000.

Type: Primary
Antigen: ZNF484
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 100-127
Host: Rabbit
Isotype: Ig
Reactivity: Human