Anti-Il2 Rat Monoclonal Antibody [clone: JES6-5H4] (PE (Phycoerythrin))
Catalog # PRSI77-204
Supplier: ProSci Inc.
Specifications
- Antibody type:Primary
- Antigen name:interleukin 2
- Antigen symbol:Il2
- Clonality:Monoclonal
- Clone:JES6-5H4
- Conjugation:PE (Phycoerythrin)
- Flow cytometry:Yes
- Host:Rat
- Isotype:IgG2b kappa
- Cross adsorption:No
- Form:liquid
- Gene ID:P04351
- Antigen synonyms:Il2
- Storage buffer:Phosphate-buffered aqueous solution, ≤0,09% Sodium azide, may contain carrier protein/stabiliser, ph 7,2.
- Storage temperature:The product should be stored undiluted at 4 °C and should be protected from prolonged exposure to light. Do not freeze.
- Shipping temperature:4 °C
- Purification:The monoclonal antibody was purified utilizing affinity chromatography and unreacted dye was removed from the product.
- Pk:0,025 mg
Specifications
About this item
The JES6-5H4 antibody specifically binds to mouse IL-2, a 17 kDa immunoregulatory cytokine and growth factor for T and B lymphocytes, NK cells, monocytes, and macrophages. IL-2 induces the cytotoxic operations of NK cells, activates B cells, and plays a role in the differentiation of T cells. JES6-5H4 seems to have a neutralizing effect on interleukin-2.
Type: Primary
Antigen: Il2
Clonality: Monoclonal
Clone: JES6-5H4
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG2b kappa
Reactivity: Mouse